Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HFE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584157
Description
Immunogen sequence: LGLSLFEALG YVDDQLFVFY DHESRRVEPR TPWVSSRISS QMWLQLSQSL KGWDHMFTVD FWTIMENHNH SKESHTLQVI LGCEMQEDNS TEGYWKYGYD GQDHLEFCPD TLDWRAAEPR AWPTKLEWE.
The protein encoded by this gene is a membrane protein that is similar to MHC class I-type proteins and associates with beta2-microglobulin. It is thought that this protein functions to regulate iron absorption by regulating the interaction of the transferrin receptor with transferrin. The iron storage disorder, hereditary haemochromatosis, is a recessive genetic disorder that results from defects in this gene. At least nine alternatively spliced variants have been described for this gene. Additional variants have been found but their full-length nature has not been determined.
Specifications
HFE | |
Polyclonal | |
Unconjugated | |
HFE | |
hemochromatosis; hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; hereditary hemochromatosis protein homolog; Hfe; HFE 1; HFE1; HH; high Fe; HLA H; HLAH; HLA-H; MGC:150812; MGC10379; MGC103790; MHC class I-like protein HFE; Mr2; MVCD7; RT1-CAFE; TFQTL2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3077 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q30201 | |
HFE | |
Recombinant protein corresponding to Human HFE. Recombinant protein control fragment (Product #RP-103158). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction