Learn More
HFE2 Mouse anti-Human, Clone: OTI3H2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809327

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
HFE2 is involved in iron metabolism. It may be a component of the signaling pathway which activates hepcidin or it may act as a modulator of hepcidin expression. It could also represent the cellular receptor for hepcidin.Specifications
HFE2 | |
Monoclonal | |
Unconjugated | |
HFE2 | |
HFE2A, HJV, JH, RGMC | |
Mouse | |
Affinity Chromatography | |
RUO | |
148738 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3H2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q6ZVN8 | |
HFE2 | |
Human recombit protein fragment corresponding to amino acids 36-263 of human HFE2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.