Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HFM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109810
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109810 Supplier Invitrogen™ Supplier No. PA5109810
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLKSNDCLFS LENLFFEKPD EVENHPDNEK SLDWFLPPAP LISEIPDTQE LEEELESHKL LGQEKRPKML TSNLKITNED TNYISLTQKF QFAFPSDKYE QDDLNLEGVG NNDLSHVAGK LTYASQKYKN HIGTEIAPEK SVPDDTKLVN FAEDKGESTS VFRKRLFKIS DNIHGSAYSN DNELDSHIGS VKIVQTEMNK GKSRNYSNSK QKFQYSANVF TANNAFSASE IGEGMFKAPS.

The protein encoded by this gene is thought to be an ATP-dependent DNA helicase and is expressed mainly in germ-line cells. Defects in this gene are a cause of premature ovarian failure 9 (POF9).
TRUSTED_SUSTAINABILITY

Specifications

Antigen HFM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HFM1
Gene Accession No. A2PYH4, D3Z4R1
Gene Alias A330009G12Rik; Gm1046; helicase; helicase-like protein HFM1; Hfm1; HFM1, ATP dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog (S. cerevisiae); Mer3; POF9; probable ATP-dependent DNA helicase HFM1; RP11-539G11.1; SEC3D1; SEC63 domain containing 1; SEC63 domain-containing protein 1; SEC63D1; Si-11; Si-11-6
Gene Symbols HFM1
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164045, 330149, 690161
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.