Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HFM1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3800 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3800 Supplier Bioss Supplier No. BS15471R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is thought to be an ATP-dependent DNA helicase and is expressed mainly in germ-line cells. Defects in this gene are a cause of premature ovarian failure 9 (POF9).

Specifications

Antigen HFM1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HFM1
Gene Accession No. A2PYH4
Gene Alias A330009G12Rik; Gm1046; helicase; helicase-like protein HFM1; Hfm1; HFM1, ATP dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog; HFM1, ATP-dependent DNA helicase homolog (S. cerevisiae); Mer3; POF9; Probable ATP-dependent DNA helicase HFM1; RP11-539G11.1; SEC3D1; SEC63 domain containing 1; SEC63 domain-containing protein 1; SEC63D1; Si-11; Si-11-6
Gene Symbols HFM1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HFM1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164045
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.