Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGD Antibody (20D12), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | HGD |
---|---|
Clone | 20D12 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
HGD Monoclonal antibody specifically detects HGD in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
HGD | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AKU, EC 1.13.11.5, HGOFLJ94126, homogentisate 1,2-dioxygenase, homogentisate 1,2-dioxygenase (homogentisate oxidase), homogentisate oxidase, Homogentisate oxygenase, Homogentisic acid oxidase, homogentisicase | |
A synthesized peptide derived from Human HGD [UniProt Q93099] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
20D12 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3081 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title