Learn More
Invitrogen™ HGD Monoclonal Antibody (1F1)

Description
Peptide Sequence: MAELKYISGF GNECSSEDPR CPGSLPEGQN NPQVCPYNLY AEQLSGSAFT CPRSTNKRSW LYRILPSVSH KPFESIDEGH VTHNWDEVDP DPNQLRWKPF EIPKASQKKV DFVSGLHTLC GAGDIKSNNG LAIHIFLCNT SMENRCFYNS DGDFLIVPQK GNLLIYTEFG KMLVQPNEIC VIQRGMRFSI DVFEETRGYI LEVYGVHLEL PDLGPIGANG LANPRDFLIP IAWYEDRQVP GGYTVINKYQ GKLFAAKQDV SPFNVVAWHG NYTPYKYNLK NFMVINSVAF DHADPSIFTV LTALRRPARS SWHLRGLPMA PWHLCLNHL.
Specifications
Specifications
| Antigen | HGD |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 1F1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HGD |
| Gene Accession No. | Q93099 |
| Gene Alias | aku; alkaptonuria; HGD; HGO; homogentisate 1, 2-dioxygenase; homogentisate 1,2-dioxygenase; homogentisate oxidase; homogentisate oxygenase; Homogentisic acid oxidase; homogentisicase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.