Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562592
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562592 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562592 Supplier Invitrogen™ Supplier No. PA562592
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AELKYISGFG NECSSEDPRC PGSLPEGQNN PQVCPYNLYA EQLSGSAFTC PRSTNKRSWL YRILPSVSHK PFESIDEGQV THNWDEVDPD PNQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 89%.

This gene encodes the enzyme homogentisate 1,2 dioxygenase. This enzyme is involved in the catabolism of the amino acids tyrosine and phenylalanine. Mutations in this gene are the cause of the autosomal recessive metabolism disorder alkaptonuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HGD
Gene Accession No. Q93099
Gene Alias aku; alkaptonuria; HGD; HGO; homogentisate 1, 2-dioxygenase; homogentisate 1,2-dioxygenase; homogentisate oxidase; homogentisate oxygenase; Homogentisic acid oxidase; homogentisicase
Gene Symbols HGD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HGD. Recombinant protein control fragment (Product #RP-103759).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.