Learn More
Invitrogen™ HGD Recombinant Rabbit Monoclonal Antibody (7O8Q1)

Description
Positive test controls include: HT-29, HepG2. Immunogen sequence: IRGHYEAKQG GFLPGGGSLH STMTPHGPDA DCFEKASKVK LAPERIADGT MAFMFESSLS LAVTKWGLKA SRCLDENYHK CWEPLKSHFT PNSRNPAEPN.
Specifications
Specifications
| Antigen | HGD |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 7O8Q1 |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HGD |
| Gene Accession No. | Q93099 |
| Gene Alias | aku; alkaptonuria; HGD; HGO; homogentisate 1, 2-dioxygenase; homogentisate 1,2-dioxygenase; homogentisate oxidase; homogentisate oxygenase; Homogentisic acid oxidase; homogentisicase |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.