Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGF Activator Antibody (101), DyLight 650 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289418C
Description
HGF Activator Monoclonal antibody specifically detects HGF Activator in Human samples. It is validated for ELISASpecifications
HGF Activator | |
Monoclonal | |
DyLight 650 | |
HGF activator, HGFA, HGFAC | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF Activator (NP_001519.1; Met 1-Ser 655). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
101 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3083 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction