Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGF Activator Monoclonal Antibody (6C2C2G11)
GREENER_CHOICE

Catalog No. PIMA529321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529321 Supplier Thermo Scientific Supplier No. MA529321
Add to Cart
Add to Cart

HGF Activator Monoclonal Antibody (6C2C2G11)

HGF Activator Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen HGF Activator
Applications ELISA
Classification Monoclonal
Clone 6C2C2G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Hgfac
Gene Accession No. Q04756
Gene Alias hepatocyte growth factor activator; Hepatocyte growth factor activator long chain; Hepatocyte growth factor activator short chain; HGF activator; HGFA; Hgfac
Gene Symbols Hgfac
Host Species Mouse
Immunogen Recombinant Human HGFA protein (Met1-Ser655).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.