Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGF Antibody (SBF5 C1.7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002696B
Description
HGF Monoclonal specifically detects HGF in Human samples. It is validated for Western Blot, ELISA.Specifications
HGF | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HGF | |
Recombinant Human HGF | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
3082 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
SBF5 C1.7 | |
Western Blot, ELISA, Immunohistochemistry | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
SBF5 C1.7 does not cross react with other members of the Skingle-serine protease superfamily such as HGF1/MSP, plasminogen/plasmin. t-PA and u-PA. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction