Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGF Monoclonal Antibody (2D2B4), NeutraKine™, Proteintech
SDP

Catalog No. 502334375 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4375 Supplier Proteintech Group Inc Supplier No. 690271IG100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lyophilized antibodies are stable for 1 year from the date of receipt if stored between (-20C) and (-80C). Upon reconstitution we recommend that the solution can be stored at(4C) for short term or at(-20C) to (-80C) for long term. Repeated freeze thaw cycles should be avoided with reconstituted products.

HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for ELISA, Neutralization

Specifications

Antigen HGF
Applications Neutralization
Classification Monoclonal
Clone 2D2B4
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene Accession No. P14210
Gene Alias DFNB39; F-TCF; HGF; HGFB; HPTA; SF
Host Species Mouse
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3082
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.