Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGF Monoclonal Antibody (OTI1C6), TrueMAB™, OriGene
SDP

Catalog No. 501681427
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1427 Supplier OriGene Supplier No. TA807185
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatocyte growth factor (HGF) is a member of the plasminogen subfamily of S1 peptidases. The inactive polypeptide formed upon translation undergoes cleavage to produce a 69 kDa alpha-chain and 34 kDa beta-chain. A disulphide bond between the two chains makes active HGF. HGF is secreted by mesenchymal cells and stimulates mitogenesis, cell motility, and matrix invasion. It is involved in cellular growth and morphogenesis in a wide variety of tissue types. In humans, HGF is present on chromosome 7.

Specifications

Antigen HGF
Applications Western Blot
Classification Monoclonal
Clone OTI1C6
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HGF
Gene Accession No. P14210
Gene Alias DFNB39, F-TCF, HGFB, HPTA, SF
Gene Symbols HGF
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 200-494 of human HGF produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3082
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.