Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGFR/c-MET Antibody (243), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289295AF350
Description
HGFR/c-MET Monoclonal specifically detects HGFR/c-MET in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
HGFR/c-MET | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
243 | |
Flow Cytometry, ELISA | |
AUTS9, c-Met, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF receptor, HGF/SF receptor, HGFR, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human HGFR/c-MET (NP_000236; Met1-Thr932). | |
0.1 mL | |
Cancer, Cancer Stem Cells, Cell Cycle and Replication, Cellular Markers, Oncogenes, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
4233 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction