Learn More
Invitrogen™ HGH1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total HGH1 protein.
Specifications
Specifications
| Antigen | HGH1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HGH1 |
| Gene Accession No. | Q9BTY7 |
| Gene Alias | brain protein 16; brain protein 16-like; Brp16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; Fam203a; FAM203B; family with sequence similarity 203, member A; family with sequence similarity 203, member B; Hgh1; HGH1 homolog; MNCb-5873; protein FAM203A; protein FAM203B; protein HGH1 homolog |
| Gene Symbols | HGH1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.