Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559987 Supplier Invitrogen™ Supplier No. PA559987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVEAIMLLTA TAPGRQQVRD QGAYLILREL HSWEPEPDVR TACEKLIQVL IGDEPERGME NLLEVQVPED VEQQLQQLDC REQEQLEREL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 82%.

Hgh1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HGH1
Gene Accession No. Q9BTY7
Gene Alias brain protein 16; brain protein 16-like; Brp16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; Fam203a; FAM203B; family with sequence similarity 203, member A; family with sequence similarity 203, member B; Hgh1; HGH1 homolog; MNCb-5873; protein FAM203A; protein FAM203B; protein HGH1 homolog
Gene Symbols HGH1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C8orf30A. Recombinant protein control fragment (Product #RP-104337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.