Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGK/MAP4K4 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074630AF594
Description
HGK/MAP4K4 Polyclonal antibody specifically detects HGK/MAP4K4 in Human samples. It is validated for Western BlotSpecifications
HGK/MAP4K4 | |
Polyclonal | |
Western Blot | |
EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111 | |
The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human MAPK/ERK kinase kinase kinase 4 (hepatocyte progenitor kinase-like/germinal center kinase-like kinase) using the numbering given in entry AAO32626.1 (GeneID 94 | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
9448 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction