Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10055249
Description
HGS Polyclonal specifically detects HGS in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
HGS | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
FLJ39878, hepatocyte growth factor-regulated tyrosine kinase substrate, Hrs, HRSFLJ11421, human growth factor-regulated tyrosine kinase substrate, Protein pp110, Vps27, ZFYVE8 | |
Rabbit | |
Affinity Purified | |
RUO | |
9146 | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
O14964 | |
HGS | |
The immunogen recognized by this antibody maps to a region between residue 727 and the C-terminus (resude 777) of human growth factor-regulated tyrosine kinase substrate using the numbering given in entry NP_004703.1 (GeneID 9146). | |
0.1 mL | |
Primary | |
HRS . | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction