Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGS Antibody (OTI3H7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271418F
Description
HGS Monoclonal specifically detects HGS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HGS | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HGS | |
Full length recombinant protein of human HGS (NP_004703) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunocytochemistry, Immunofluorescence | |
OTI3H7 | |
Immunocytochemistry/Immunofluorescence | |
FLJ39878, hepatocyte growth factor-regulated tyrosine kinase substrate, Hrs, HRSFLJ11421, human growth factor-regulated tyrosine kinase substrate, Protein pp110, Vps27, ZFYVE8 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
9146.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction