Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HGS Antibody (OTI3H7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245734
Description
Ensure accurate, reproducible results in Western Blot, Immunofluorescence
HGS Monoclonal specifically detects HGS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
HGS | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:4000, Immunofluorescence 1:100 | |
FLJ39878, hepatocyte growth factor-regulated tyrosine kinase substrate, Hrs, HRSFLJ11421, human growth factor-regulated tyrosine kinase substrate, Protein pp110, Vps27, ZFYVE8 | |
Mouse | |
86 kDa | |
0.1 mL | |
Cell Cycle and Replication, Signal Transduction | |
9146 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunofluorescence | |
OTI3H7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
HGS | |
Full length human recombinant protein of human HGS(NP_004703) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction