Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595931 Supplier Invitrogen™ Supplier No. PA595931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EQQKQTVQMR AQMPAFPLPY AQLQAMPAAG GVLYQPSGPA SFPSTFSPAG SVEGSPMHGV YMSQPAPAAG PYPSMPSTAA DPSMVSAYMY PAGATGAQAA PQAQAGPTAS PAYSSYQPTP TAGYQNVASQ APQSLPAISQ PPQSSTMGYM GSQSVSMGYQ PYNMQNLMTT LPSQDASLPP QQPYIAGQQP MYQQMAPSGG PPQQQPPVAQ QPQAQGPPAQ GSEAQLISFD; Positive Samples: THP-1, OVCAR3, MCF7, HepG2, Mouse testis, Rat testis; Cellular Location: Cytoplasm, Cytoplasmic side, Early endosome membrane, Endosome, Peripheral membrane protein, multivesicular body membrane.

HGS is involved in intracellular signal transduction mediated by cytokines and growth factors. When associated with STAM, it suppresses DNA signaling upon stimulation by IL-2 and GM-CSF. It could be a direct effector of PI3-kinase in vesicular pathway via early endosomes and may regulate trafficking to early and late endosomes by recruiting clathrin. HGS may concentrate ubiquitinated receptors within clathrin-coated regions. It is involved in down-regulation of receptor tyrosine kinase via multivesicular body (MVBs) when complexed with STAM. This complex binds ubiquitin and acts as sorting machinery that recognizes ubiquitinated receptors and transfers them to further sequential lysosomal sorting/trafficking processes. HGS may contribute to the efficient recruitment of SMADs to the activin receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGS
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene HGS
Gene Accession No. O14964, Q99LI8, Q9JJ50
Gene Alias hepatocyte growth factor-regulated tyrosine kinase substrate; HGF-regulated tyrosine kinase substrate; HGNC:4897; Hgr; Hgs; Hrs; Hrs2; human growth factor-regulated tyrosine kinase substrate; protein pp110; SNAP-25-interacting protein Hrs-2; ZFYVE8
Gene Symbols HGS
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 548-777 of human HGS (NP_0047031).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15239, 56084, 9146
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.