Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HGSNAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556197
Description
Immunogen sequence: QALLLIHNEL LWTNLTVYWK SECCYHCLFQ VLVNVPQSPK AGKPSAAAAS VSTQHGSILQ LNDTLEEKEV CRLEYRFGEF GNYSLLVKNI HNGVSEIACD LAVN Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 23%.
This gene encodes a lysosomal acetyltransferase, which is one of several enzymes involved in the lysosomal degradation of heparin sulfate. Mutations in this gene are associated with Sanfilippo syndrome C, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate.
Specifications
HGSNAT | |
Polyclonal | |
Unconjugated | |
HGSNAT | |
Heparan-alpha-glucosaminide N-acetyltransferase; HGNAT; HGSNAT; MPS3C; RP73; TMEM76; transmembrane protein 76 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
138050 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q68CP4 | |
HGSNAT | |
Recombinant protein corresponding to Human HGSNAT. Recombinant protein control fragment (Product #RP-90544). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction