Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HGSNAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545749 Supplier Invitrogen™ Supplier No. PA545749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RALAALLLAA SVLSAALLAP GGSSGRDAQA APPRDLDKKR HAELKMDQAL Sequence homology: Dog: 86%; Horse: 82%; Human: 100%.

This gene encodes a lysosomal acetyltransferase, which is one of several enzymes involved in the lysosomal degradation of heparin sulfate. Mutations in this gene are associated with Sanfilippo syndrome C, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGSNAT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HGSNAT
Gene Accession No. Q68CP4
Gene Alias Heparan-alpha-glucosaminide N-acetyltransferase; HGNAT; HGSNAT; MPS3C; RP73; TMEM76; transmembrane protein 76
Gene Symbols HGSNAT
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HGSNAT (aa 34-83).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138050
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.