Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ HGT-1 Human Gastric Cancer Cell Line

Catalog No. scc199mi Shop All MilliporeSigma Products

HGT-1 human gastric cancer cell line is used for characterization and tumorigenic studies.

  • Derived from a poorly-differentiated adenocarcinoma and used as a model for studies of gastric cancer and mechanisms of gastric secretion
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen, Cells can be cultured at least 10 passages after initial thawing without significantly affecting cell marker expression and functionality
Cell Type Cancer Cells
For Use With (Application) Cell Culture, Characterization and Tumorigenic Studies
No. of Cells ≥ 1 x 106 Cells/Vial

For research purposes only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.