Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HGV Polyprotein Rabbit anti-Virus, Polyclonal, Bioss
SDP

Catalog No. 501980221
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0221 Supplier Bioss Supplier No. BS0338R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis G virus is from a group of viruses called GB viruses (GB virus A, GB virus B and GB virus C) and is also designated as GB virus C (GBV C). HGV is a positive stranded RNA virus, with a genome of 10 KB and appears, on the basis of genome length and organisation, to be a member of the Flaviviridae and distinct from the hepatitis C virus (HCV). HGV infection appears to be transmitted following receipt of blood from HGV RNA positive donors, however, these patients have only moderate liver enzyme elevations and do not develop chronic liver disease.

Specifications

Antigen HGV Polyprotein
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Hepatitis G Virus Polyprotein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.