Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HHAT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA525417
Description
HHAT Polyclonal Antibody for Western Blot, IHC (P), Flow
Skinny hedgehog' (SKI1) encodes an enzyme that acts within the secretory pathway to catalyze amino-terminal palmitoylation of 'hedgehog' (see MIM 600725).
Specifications
HHAT | |
Polyclonal | |
Unconjugated | |
HHAT | |
Hedgehog acyltransferase; HHAT; MART2; MART-2; melanoma antigen recognized by T cells 2; melanoma antigen recognized by T-cells 2; Protein-cysteine N-palmitoyltransferase HHAT; SKI1; Skinny hedgehog protein 1; Skn | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
55733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5VTY9 | |
HHAT | |
KLH conjugated synthetic peptide between 30-59 amino acids from the N-terminal region of human HHAT. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction