Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHATL/GUP1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS12315R
Description
The Protein-cysteine N-palmitoyltransferase HHAT-like protein (HHATL), also known at the mammalian glycerol uptake/transporter 1 (Gup1) is a homolog of the S. cerevisiae Gup1. It is an endoplasmic reticulum (ER) membrane protein that co-localizes with the mammalian hedgehog acyltransferse Skn and interferes with its N-terminal palmitoylation of Sonic hedgehog (SHH), suggesting that HHATL acts as a negative regulator for N-terminal palmitoylation of SHH. HHATL is highly expressed in normal skin, but becomes down-regulated in skin squamous cell carcinoma, suggesting it may play a role in its development.Specifications
| HHATL/GUP1 | |
| Polyclonal | |
| Unconjugated | |
| HHATL | |
| 1110011D13Rik; C3orf3; Glycerol uptake/transporter homolog; GUP1; GUP1 glycerol uptake/transporter homolog; Gup1, glycerol uptake/transporter homolog; hedgehog acyltransferase-like; Hedgehog acyltransferase-like protein; Hhatl; KIAA1173; MBOAT3; membrane bound O-acyltransferase domain containing 3; MSTP002; OACT3; Protein-cysteine N-palmitoyltransferase HHAT-like protein; RGD1311911 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 57467 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q9HCP6 | |
| HHATL | |
| KLH conjugated synthetic peptide derived from human HHATL/GUP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction