Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HHATL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109811
Description
Immunogen sequence: WQSGFVTGTF DLQEVLFHGG SSFTVLRCTS FALESCAHPD RHYSLADLLK YNFYLPFFFF GPIMTFDRFH AQVSQVEPVR REGELW.
HHATL (Hedgehog Acyltransferase-Like) is a gene that codes for a protein involved in the regulation of Hedgehog signaling pathways. HHATL is located on chromosome 7q36.3. Structurally, the protein encoded by this gene is characterized by an acyltransferase domain, which plays a crucial role in the N-palmitoylation of the Sonic Hedgehog (Shh) protein, a process essential for its proper signaling activity and distribution. Functionally, HHATL has been implicated in various biological processes, including amelioration of endoplasmic reticulum stress through autophagy. The gene is expressed in several tissues, with notable high expression in the heart, suggesting a potential role in cardiac development and function. Dysfunction in HHATL could lead to anomalies in Hedgehog signaling, impacting various developmental and cellular processes.
Specifications
HHATL | |
Polyclonal | |
Unconjugated | |
HHATL | |
1110011D13Rik; C3orf3; Glycerol uptake/transporter homolog; GUP1; GUP1 glycerol uptake/transporter homolog; Gup1, glycerol uptake/transporter homolog; hedgehog acyltransferase-like; hedgehog acyltransferase-like protein; HHATL; KIAA1173; MBOAT3; membrane bound O-acyltransferase domain containing 3; MSTP002; OACT3; Protein-cysteine N-palmitoyltransferase HHAT-like protein; RGD1311911 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
301073, 57467 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.55 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9HCP6 | |
HHATL | |
Recombinant fusion protein containing a sequence corresponding to amino acids 160-245 of human HHATL (NP_065758.3). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction