Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHATL Rabbit anti-Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096088 20 μL
NB096087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096088 Supplier Novus Biologicals Supplier No. NBP30474520UL

Rabbit Polyclonal Antibody

HHATL Polyclonal specifically detects HHATL in Rat samples. It is validated for Western Blot.

Specifications

Antigen HHATL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias C3orf3, chromosome 3 open reading frame 3, glycerol uptake/transporter homolog, GUP1, hedgehog acyltransferase-like, Hedgehog acyltransferase-like protein, KIAA1173GUP1, glycerol uptake/transporter homolog (yeast), MBOAT3, membrane bound O-acyltransferase domain containing 3, protein-cysteine N-palmitoyltransferase HHAT-like protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-245 of human HHATL (NP_065758.3).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57467
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.