Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHBrk1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986347
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6347 Supplier Bioss Supplier No. BS5124R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C3orf10 is involved in regulation of actin and microtubule organization. It is a part of a WAVE complex that activates the Arp2/3 complex.

Specifications

Antigen hHBrk1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BRK1
Gene Accession No. Q8WUW1
Gene Alias 6720456B07Rik; AW011779; BOS_20736; BRICK1, SCAR/WAVE actin nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit; BRICK1, SCAR/WAVE actin-nucleating complex subunit, homolog; brk1; C22H3orf10; C3orf10; haematopoietic stem/progenitor cell protein 300; hHBrk1; HSPC300; LOC100058184; LOC100522899; MDS027; Probable protein BRICK1; probable protein BRICK1-like; Protein BRICK1; zgc:86903
Gene Symbols BRK1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human hHBrk1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55845
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.