Learn More
Invitrogen™ HHCM Polyclonal Antibody

Description
HHCM Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | HHCM |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | HHCM |
| Gene Accession No. | 0 |
| Gene Alias | Hepatocellular carcinoma protein HHCM |
| Gene Symbols | HHCM |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.