Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHEX Antibody (2018B) [PerCP], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB83771C
Description
HHEX Monoclonal antibody specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
HHEX | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
2018B | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
E. coli-derived recombinant human HHEX, Thr111-Gly270, Accession # Q03014 | |
0.1 mL | |
Developmental Biology | |
3087 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction