Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHEX Antibody (OTI3E6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270890F
Description
HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
HHEX | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
HHEX | |
Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of human HHEX. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunocytochemistry, Immunofluorescence | |
OTI3E6 | |
Immunocytochemistry/Immunofluorescence | |
hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
Mouse | |
Affinity Purified | |
Developmental Biology | |
3087.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction