Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Hexosaminidase A/HEXA Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB62371SP
This item is not returnable.
View return policy
Description
Hexosaminidase A/HEXA Monoclonal specifically detects Hexosaminidase A/HEXA in Human samples. It is validated for Neutralization.Specifications
Hexosaminidase A/HEXA | |
Monoclonal | |
LYOPH | |
Neutralization 3.0 ug/mL | |
P06865 | |
HEXA | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Hexosaminidase A/HEXA Met1-Thr529 Accession # P06865 | |
25 μg | |
Primary | |
Detects human Hexosaminidase A/HEXA in direct ELISAs. | |
Human | |
Purified |
Neutralization | |
714712 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta-hexosaminidase subunit alpha, Beta-N-acetylhexosaminidase subunit alpha, EC 3.2.1, EC 3.2.1.52, HEXA, hexosaminidase A (alpha polypeptide), Hexosaminidase subunit A, MGC99608, N-acetyl-beta-glucosaminidase subunit alpha, TSD | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3073 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction