Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHIP Recombinant Rabbit Monoclonal Antibody (HL1737)
GREENER_CHOICE

Catalog No. PIMA547163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547163 Supplier Invitrogen™ Supplier No. MA547163
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signaling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHIP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1737
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene HHIP
Gene Accession No. Q7TN16, Q96QV1
Gene Alias hedgehog interacting protein; Hedgehog-interacting protein; Hedgehog-interacting protein-like; Hhip; Hhip1; HIP; RGD1564108; UNQ5825/PRO19644
Gene Symbols HHIP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human HHIP. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15245, 64399
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.