Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115355
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115355 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115355 Supplier Invitrogen™ Supplier No. PA5115355
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Hhip.

This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signaling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHIP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene HHIP
Gene Accession No. Q7TN16, Q96QV1
Gene Alias hedgehog interacting protein; Hedgehog-interacting protein; Hedgehog-interacting protein-like; Hhip; Hhip1; HIP; RGD1564108; UNQ5825/PRO19644
Gene Symbols HHIP
Host Species Rabbit
Immunogen A synthesized peptide derived from human HHIP(Accession Q96QV1), corresponding to amino acid residues E640-S690.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15245, 291936, 64399
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.