Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHIPL1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA562678
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562678 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562678 Supplier Thermo Scientific Supplier No. PA562678

Rabbit Polyclonal Antibody

Immunogen sequence: GRLMSLQENP GTGQWQYSEI CMGHGQTCEF PGLINNYYPY IISFG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

This gene encodes a protein that belongs to the glucose/sorbosone dehydrogenase family. The encoded protein also contains a domain that binds folate and reduced folic acid derivatives.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHIPL1
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HHIPL1
Gene Accession No. Q96JK4
Gene Alias 1600002O04Rik; AK129450; ARAR9245; hedgehog interacting protein-like 1; HHIP like 1; HHIP2; HHIPL1; HHIP-like 1; HHIP-like protein 1; KIAA1822; LOW QUALITY PROTEIN: HHIP-like protein 1; mKIAA1822; RGD1311698; UNQ9245; UNQ9245/PRO34761
Gene Symbols HHIPL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HHIPL1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.