Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHIPL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983804
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3804 Supplier Bioss Supplier No. BS15476R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein that belongs to the glucose/sorbosone dehydrogenase family. The encoded protein also contains a domain that binds folate and reduced folic acid derivatives.

Specifications

Antigen HHIPL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HHIPL1
Gene Accession No. Q96JK4
Gene Alias 1600002O04Rik; AK129450; ARAR9245; hedgehog interacting protein-like 1; HHIP like 1; Hhip2; HHIPL1; HHIP-like 1; HHIP-like protein 1; Kiaa1822; LOW QUALITY PROTEIN: HHIP-like protein 1; mKIAA1822; RGD1311698; UNQ9245; UNQ9245/PRO34761
Gene Symbols HHIPL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HHIPL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84439
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.