Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHIPL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563765 Supplier Invitrogen™ Supplier No. PA563765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LCSDYCSAFH SNCHSAISLL TNDRGLQESH GRDGTRFCHL LDLPDKDYCF PNVLRNDYLN RHLGMVAQDP QGC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

HHIPL2 gene ontology annotations related to this gene include catalytic activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHIPL2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HHIPL2
Gene Accession No. Q6UWX4
Gene Alias hedgehog interacting protein-like 2; HHIP like 2; HHIP3; HHIPL2; HHIP-like 2; HHIP-like protein 2; KIAA1822L; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; UNQ841/PRO1779
Gene Symbols HHIPL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HHIPL2. Recombinant protein control fragment (Product #RP-103699).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.