Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHIPL2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983805
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3805 Supplier Bioss Supplier No. BS15477R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HHIPL2 gene ontology annotations related to this gene include catalytic activity.

Specifications

Antigen HHIPL2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HHIPL2
Gene Accession No. Q6UWX4
Gene Alias hedgehog interacting protein-like 2; HHIP like 2; HHIP3; HHIPL2; HHIP-like 2; HHIP-like protein 2; KIAA1822L; UNQ841/PRO1779
Gene Symbols HHIPL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HHIPL2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79802
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.