Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHMJG Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303589100UL
Description
HHMJG Polyclonal specifically detects HHMJG in Human, Mouse samples. It is validated for Western Blot.Specifications
HHMJG | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
8690 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
erky (mouse) homolog-like, HHMJGHuman homolog of mouse jerky gene protein, jerky homolog-like (mouse), jerky protein homolog-like, MGC129608 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 225-524 of human HHMJG (NP_003763.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction