Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hHR23b Antibody (OTI7G10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271269F
Description
hHR23b Monoclonal specifically detects hHR23b in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
hHR23b | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
RAD23B | |
Recombinant protein fragment corresponding to amino acids 1-253 of human hHR23b (NP_002865) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI7G10 | |
Immunohistochemistry | |
HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors | |
5887.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction