Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hHR23b Antibody (SAIC-28A-29), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320096IR
Description
hHR23b Monoclonal antibody specifically detects hHR23b in Human samples. It is validated for Western BlotSpecifications
hHR23b | |
Monoclonal | |
DyLight 755 | |
HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein | |
Peptide "e;ILNDDTALK"e;, derived from the UV excision repair protein RAD23 homolog B, conjugated to KLH. | |
0.1 mL | |
Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors | |
5887 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot | |
SAIC-28A-29 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction