Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHV-1 TK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA567984

Description
The Thymidine kinase catalyzes the transfer of the gamma-phospho group of ATP to thymidine to generate dTMP in the salvage pathway of pyrimidine synthesis. The dTMP serves as a substrate for DNA polymerase during viral DNA replication. The thymidine kinase allows the virus to be reactivated and to grow in non-proliferative cells lacking a high concentration of phosphorylated nucleic acid precursors. It also phosphorylates and thereby activates certain drugs like acyclovir (ACV), valacyclovir, and famciclovir to a toxic form, that leads to successful suppression of the infection, while the uninfected cell does not have this ability because it lacks TK. Mutations in thymidine kinase may induce HSV resistance to antiviral therapies in immunocompromised patients. The most frequently observed resistant strains are unable to express TK and are avirulent in animal models of disease. Resistance may be acquired less frequently by selecting variants which no longer recognize ACV or ACV triphosphate as substrates but which retain normal functions.
Specifications
HHV-1 TK | |
Polyclonal | |
Unconjugated | |
Rabbit | |
affinity chromatography | |
RUO | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Synthesized peptide derived from human KITH_HHV11. | |
100 μL | |
Primary | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction