Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHV-1 US3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567986 Supplier Invitrogen™ Supplier No. PA567986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HHV-1 US3 Polyclonal Antibody for Western Blot

US3 is a multifunctional serine/threonine kinase that plays a role in several processes including egress of virus particles from the nucleus, modulation of the actin cytoskeleton and inhibition of apoptosis. It phosphorylates UL31 and UL34, two critical regulators of capsid budding from nucleus to endoplasmic reticulum, thereby facilitating virion egress. US3 modulates and redistributes host components of the nuclear envelope, including LMNA, emerin/EMD and the nuclear matrix protein MATR3. It also phosphorylates envelope glycoprotein B (gB), probably to direct it to the cell surface, and promotes virus intracellular spread by restructuring host cell cytoskeleton. US3 blocks host apoptosis to extend cell survival and allow efficient viral replication. Promotes viral gene expression by phosphorylating host HDAC2 to reduce viral genome silencing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHV-1 US3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Synthesized peptide derived from human KR1_HHV11.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.