Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHV-6 Envelope Monoclonal Antibody (F59G)
GREENER_CHOICE

Catalog No. MA17044
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17044 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17044 Supplier Invitrogen™ Supplier No. MA17044
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-7044 detects the human Herpes Virus, type 6. This antibody is specific for the 60/110 kDa envelope glycoprotein of HHV6. MA1-7044 has been successfully used in Western blot, immunoprecipitaion, immunofluorescence and immunohistochemistry procedures. HIER is recommended for use in IHC on paraffin embedded sections. The MA1-7044 immunogen is Human Herpes Virus type 6 lysate.

TRUSTED_SUSTAINABILITY

Specifications

Antigen HHV-6 Envelope
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F59G
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HHV-6; HHV-6 Envelope
Host Species Mouse
Immunogen Human Herpes Virus type 6 lysate.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.