Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HHV-6A U69 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567991 Supplier Invitrogen™ Supplier No. PA567991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HHV-6A U69 Polyclonal Antibody for Western Blot

U69 phosphorylates the antiviral nucleoside analog ganciclovir.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHV-6A U69
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Synthesized peptide derived from human GCVK_HHV6U.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.