Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHV DNA polymerase catalytic subunit Rabbit anti-Virus, Polyclonal, Bioss
SDP

Catalog No. 501980905
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0905 Supplier Bioss Supplier No. BS10489R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Replicates viral genomic DNA. The replication complex is composed of six viral proteins: the DNA polymerase, processivity factor, primase, primase-associated factor, helicase, and ssDNA-binding protein. Additionally, the polymerase contains an intrinsic ribonuclease H (RNase H) activity that specifically degrades RNA/DNA heteroduplexes or duplex DNA substrates in the 5′ to 3′ direction. Therefore, it can catalyze the excision of the RNA primers that initiate the synthesis of Okazaki fragments at a replication fork during viral DNA replication.

Specifications

Antigen HHV DNA polymerase catalytic subunit
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Human herpesvirus 1 DNA polymerase catalytic subunit.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.