Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HHV14 UL38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572160
Description
HHV14 UL38 Polyclonal Antibody for Western Blot
Structural component of the T=16 icosahedral capsid. The capsid is composed of pentamers and hexamers of VP5, which are linked together by heterotrimers called triplex. These triplex are formed by a single molecule of VP19C and two copies of VP23 which bridge major capsid protein VP5 multimers together. Triplexes occupy the local threeflod positions between capsid hexamers and pentamers.
Specifications
| HHV14 UL38 | |
| Polyclonal | |
| Unconjugated | |
| TRX1 | |
| UL38; Virion protein UL38 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| 0.54 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P17586 | |
| TRX1 | |
| KLH conjugated synthetic peptide between 387-414 amino acids from the C-terminal region of human HHV14 UL38. | |
| 400 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction