Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HHV8-ORF74 Antibody (462510), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5264W
Description
HHV8-ORF74 Monoclonal antibody specifically detects HHV8-ORF74 in Virus samples. It is validated for Flow Cytometry, CyTOFSpecifications
HHV8-ORF74 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
NS0 mouse myeloma cell line transfected with viral HHV8-ORF74, Met1-Thr342, Accession # AAB51506 | |
0.1 mL | |
Primary | |
Virus | |
Purified |
Flow Cytometry, CyTOF | |
462510 | |
Flow Cytometry, CyTOF-ready | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4961460 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction